Gene Transcripts
- Gene Symbol
- FAM219A
- Entrez Gene ID
- 203259
- Gene Name
- family with sequence similarity 219 member A
- Gene Aliases
- C9orf25
- Chromosome Location
- Chr. 9: 34398184 - 34458610 on Build GRCh38
- UniGene ID
- Hs.493771
- Species
- Homo sapiens
- Gene Symbol
- C9orf24
- Entrez Gene ID
- 84688
- Gene Name
- chromosome 9 open reading frame 24
- Gene Aliases
- CBE1, NYD-SP22, SMRP1, bA573M23.4
- Chromosome Location
- Chr. 9: 34379019 - 34400833 on Build GRCh38
- UniGene ID
- Hs.50334
- Species
- Homo sapiens
View Full Product Details