Gene Transcripts

Gene Symbol
BEST4
Entrez Gene ID
266675
Gene Name
bestrophin 4
Gene Aliases
VMD2L2
Chromosome Location
Chr. 1: 44781836 - 44792828 on Build GRCh38
UniGene ID
Hs.302513
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153274.2 NP_695006.1 9 1588
XM_017001022.1 XP_016856511.1 11 2939
GenBank mRNA AF440757.1 - 9 1539

View Full Product Details