Gene Transcripts

Gene Symbol
ZYX
Entrez Gene ID
7791
Gene Name
zyxin
Gene Aliases
ESP-2, HED-2
Chromosome Location
Chr. 7: 143381174 - 143391111 on Build GRCh38
UniGene ID
Hs.490415
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010972.1 NP_001010972.1 4 576
NM_003461.4 NP_003452.1 4 579
XM_017012587.1 XP_016868076.1 4 669
GenBank mRNA AK299005.1 - 3 295
AK302309.1 - 3 387
AK316227.1 - 3 271
BC002323.2 - 4 451
BC008743.2 - 4 543 3345843
BC009360.2 - 4 488 4301542
BC010031.2 - 4 476 3506023
BC017183.2 - 3 105
BU844538.1 - 545
CR457431.1 - 3 421
U15158.1 - 2 184
X94991.1 - 3 431
X95735.1 - 4 492

View Full Product Details