Gene Transcripts

Gene Symbol
ACBD4
Entrez Gene ID
79777
Gene Name
acyl-CoA binding domain containing 4
Gene Aliases
HMFT0700
Chromosome Location
Chr. 17: 45132600 - 45144181 on Build GRCh38
UniGene ID
Hs.110298
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135705.2 NP_001129177.1 10 1627
NM_001135706.2 NP_001129178.1 10 1665
NM_001135707.2 NP_001129179.1 10 2477
NM_001321352.1 NP_001308281.1 12 1789
NM_001321353.1 NP_001308282.1 12 1786
NM_024722.3 NP_078998.1 9 1781
XM_006722085.2 XP_006722148.1 11 2000
XM_017025084.1 XP_016880573.1 11 2056
XM_017025085.1 XP_016880574.1 12 1807
XM_017025086.1 XP_016880575.1 13 1956
XM_017025087.1 XP_016880576.1 11 2038
XM_017025088.1 XP_016880577.1 11 2035
XM_017025090.1 XP_016880579.1 11 2018
XM_017025091.1 XP_016880580.1 11 1997
XM_017025092.1 XP_016880581.1 12 2122
XM_017025094.1 XP_016880583.1 9 2645
XM_017025095.1 XP_016880584.1 10 1916
XM_017025096.1 XP_016880585.1 10 1878
XM_017025097.1 XP_016880586.1 10 1895
XM_017025098.1 XP_016880587.1 11 1646
XM_017025099.1 XP_016880588.1 10 1857
GenBank mRNA AB073387.1 - 10 1577
AK023384.1 - 9 1741
BC029164.1 - 10 1566 5188183
BC041143.1 - 12 1758 5769682

View Full Product Details