Gene Transcripts

Gene Symbol
ASF1B
Entrez Gene ID
55723
Gene Name
anti-silencing function 1B histone chaperone
Gene Aliases
CIA-II
Chromosome Location
Chr. 19: 14119509 - 14136628 on Build GRCh38
UniGene ID
Hs.26516
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018154.2 NP_060624.1 4 1398
GenBank mRNA AK001288.1 - 4 1357
AK001466.1 - 4 1389
AK223080.1 - 4 1352
BC007726.2 - 4 1322 4303318
BC010014.2 - 4 1332 4304756
BC036521.1 - 4 1362 5270216

View Full Product Details