Gene Transcripts

Gene Symbol
ACER3
Entrez Gene ID
55331
Gene Name
alkaline ceramidase 3
Gene Aliases
APHC, PHCA
Chromosome Location
Chr. 11: 76860873 - 77026797 on Build GRCh38
UniGene ID
Hs.23862
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300953.1 NP_001287882.1 10 1391
NM_001300954.1 NP_001287883.1 11 1573
NM_001300955.1 NP_001287884.1 10 1449
NM_018367.6 NP_060837.3 11 1502
XM_011545151.2 XP_011543453.1 10 1366
XM_011545152.2 XP_011543454.1 11 1438
GenBank mRNA AF214454.1 - 11 1456
AF327353.1 - 11 1467
AK002100.1 - 13 1633
BC063034.1 - 12 1462

View Full Product Details