Gene Transcripts

Gene Symbol
ADGRG6
Entrez Gene ID
57211
Gene Name
adhesion G protein-coupled receptor G6
Gene Aliases
APG1, DREG, GPR126, LCCS9, PS1TP2, VIGR
Chromosome Location
Chr. 6: 142301919 - 142446266 on Build GRCh38
UniGene ID
Hs.743302
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001032394.2 NP_001027566.1 3 690
NM_001032395.2 NP_001027567.1 3 690
NM_020455.5 NP_065188.4 3 690
NM_198569.2 NP_940971.1 3 690
XM_005267061.3 XP_005267118.1 3 591
XM_006715516.2 XP_006715579.1 3 591
XM_006715517.2 XP_006715580.1 3 391
XM_006715518.2 XP_006715581.1 3 591
XM_011535964.1 XP_011534266.1 3 394
XM_017011085.1 XP_016866574.1 3 591
GenBank mRNA AB183546.1 - 3 680
AB183547.1 - 3 680
AB183548.1 - 3 680
AB183549.1 - 3 680
AF216967.1 - 3 695
AK092519.1 - 1 31
AK122617.1 - 3 579
AK307543.1 - 3 588
BC075798.1 - 3 750 30333185
BX640873.1 - 3 296
BX640971.1 - 3 616
BX648315.1 - 3 403

View Full Product Details