Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020824.3 | NP_065875.3 | 6 | 902 | |
XM_005252542.3 | XP_005252599.1 | 6 | 970 | ||
XM_005252544.4 | XP_005252601.1 | 6 | 2655 | ||
XM_011519602.2 | XP_011517904.1 | 6 | 2655 | ||
XM_011519603.2 | XP_011517905.1 | 6 | 1062 | ||
XM_011519604.2 | XP_011517906.1 | 6 | 878 | ||
XM_011519605.2 | XP_011517907.1 | 6 | 2655 | ||
XM_011519606.2 | XP_011517908.1 | 3 | 2520 | ||
XM_011519607.2 | XP_011517909.1 | 6 | 2655 | ||
XM_017016459.1 | XP_016871948.1 | 6 | 1062 | ||
XM_017016460.1 | XP_016871949.1 | 6 | 1062 | ||
XM_017016461.1 | XP_016871950.1 | 6 | 873 | ||
GenBank mRNA | AB037845.2 | - | 5 | 375 | |
AF480466.1 | - | 6 | 849 | ||
AK074589.1 | - | 1108 | |||
AK297278.1 | - | 1 | 48 | ||
AK307604.1 | - | 6 | 879 | ||
BC118915.2 | - | 5 | 506 | 40071194 |