Gene Transcripts

Gene Symbol
EPB41L5
Entrez Gene ID
57669
Gene Name
erythrocyte membrane protein band 4.1 like 5
Gene Aliases
BE37, YMO1, YRT
Chromosome Location
Chr. 2: 120013028 - 120179121 on Build GRCh38
UniGene ID
Hs.369232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184937.1 NP_001171866.1 24 3694
NM_020909.3 NP_065960.2 25 3825
XM_006712651.1 XP_006712714.1 25 3826
XM_006712652.1 XP_006712715.1 25 4141
XM_006712653.1 XP_006712716.1 25 3846
XM_011511529.2 XP_011509831.1 25 3802
XM_017004567.1 XP_016860056.1 25 3682
GenBank mRNA AB046768.1 - 18 3071
BC054508.1 - 25 3760 5502663

View Full Product Details