Gene Transcripts
- Gene Symbol
- PRKX
- Entrez Gene ID
- 5613
- Gene Name
- protein kinase, X-linked
- Gene Aliases
- PKX1
- Chromosome Location
- Chr. X: 3604343 - 3713634 on Build GRCh38
- UniGene ID
- Hs.390788
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005044.4 |
NP_005035.1
|
9 |
5633 |
|
GenBank mRNA |
X85545.1 |
- |
9 |
5599 |
|
View Full Product Details