Gene Transcripts

Gene Symbol
ANKRD1
Entrez Gene ID
27063
Gene Name
ankyrin repeat domain 1
Gene Aliases
ALRP, C-193, CARP, CVARP, MCARP, bA320F15.2
Chromosome Location
Chr. 10: 90912100 - 90921275 on Build GRCh38
UniGene ID
Hs.448589
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014391.2 NP_055206.2 9 1223
GenBank mRNA AY903446.1 - 9 992
BC018667.1 - 9 1039 4245591
X83703.1 - 9 1224

View Full Product Details