Gene Transcripts

Gene Symbol
SCAPER
Entrez Gene ID
49855
Gene Name
S-phase cyclin A associated protein in the ER
Gene Aliases
MSTP063, ZNF291, Zfp291
Chromosome Location
Chr. 15: 76348185 - 76905455 on Build GRCh38
UniGene ID
Hs.458986
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020843.2 NP_065894.2 2 148
XM_005254417.2 XP_005254474.1 3 305
XM_005254419.2 XP_005254476.1 3 305
XM_011521650.1 XP_011519952.1 4 390
XM_017022268.1 XP_016877757.1 3 253
XM_017022269.1 XP_016877758.1 3 303
XM_017022270.1 XP_016877759.1 4 461
XM_017022272.1 XP_016877761.1 3 305
XM_017022275.1 XP_016877764.1 3 303
XM_017022277.1 XP_016877766.1 3 303
XM_017022283.1 XP_016877772.1 3 305
GenBank mRNA AF242528.1 - 2 123
AK056095.1 - 3 294
AK056515.1 - 4 365
BC070197.1 - 3 165
BC107415.1 - 3 189

View Full Product Details