Gene Transcripts

Gene Symbol
ABTB2
Entrez Gene ID
25841
Gene Name
ankyrin repeat and BTB domain containing 2
Gene Aliases
ABTB2A, BTBD22
Chromosome Location
Chr. 11: 34150987 - 34358008 on Build GRCh38
UniGene ID
Hs.23361
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145804.2 NP_665803.2 17 4047
GenBank mRNA AK056863.1 - 5 1103
AK091560.1 - 14 2265
AK095632.1 - 17 3294
AL050374.1 - 2 819
BX640841.1 - 9 1748

View Full Product Details