Gene Transcripts

Gene Symbol
ADAP1
Entrez Gene ID
11033
Gene Name
ArfGAP with dual PH domains 1
Gene Aliases
CENTA1, GCS1L, p42IP4
Chromosome Location
Chr. 7: 897900 - 955407 on Build GRCh38
UniGene ID
Hs.602573
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284308.1 NP_001271237.1 11 1615
NM_001284309.1 NP_001271238.1 11 1555
NM_001284310.1 NP_001271239.1 10 1530
NM_001284311.1 NP_001271240.1 9 1383
NM_006869.3 NP_006860.1 11 1788
GenBank mRNA AB097049.1 - 11 1762
AF082324.1 - 11 1762
AK092471.1 - 10 1532
AK124979.1 - 9 1634
BC033747.1 - 11 1589 5195416

View Full Product Details