Gene Transcripts

Gene Symbol
LYPLA2
Entrez Gene ID
11313
Gene Name
lysophospholipase II
Gene Aliases
APT-2, APT2, DJ886K2.4
Chromosome Location
Chr. 1: 23790789 - 23795539 on Build GRCh38
UniGene ID
Hs.533479
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007260.2 NP_009191.1 9 725
XM_005245728.4 XP_005245785.1 9 1197
XM_017000165.1 XP_016855654.1 9 1092
GenBank mRNA AF090423.1 - 9 771
AF098668.1 - 9 725
AK024724.1 - 5 405
BC017034.2 - 9 690 3845764
BC017193.2 - 9 679 3138786

Gene Symbol
LYPLA2P2
Entrez Gene ID
388499
Gene Name
lysophospholipase II pseudogene 2
Gene Aliases
Chromosome Location
Chr. 19: 7878619 - 7880232 on Build GRCh38
UniGene ID
Hs.730360
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033250.1 1 699

View Full Product Details