Gene Transcripts

Gene Symbol
PAGE1
Entrez Gene ID
8712
Gene Name
PAGE family member 1
Gene Aliases
AL5, CT16.3, GAGE-9, GAGEB1, PAGE-1
Chromosome Location
Chr. X: 49687450 - 49696019 on Build GRCh38
UniGene ID
Hs.128231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003785.3 NP_003776.2 4 333
XM_011543998.2 XP_011542300.1 4 369
GenBank mRNA AF058989.1 - 4 329
BC004861.2 - 4 306 3829941
BU941413.1 - 4 335
CR542084.1 - 3 200

View Full Product Details