Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001160036.1 | NP_001153508.1 | 12 | 5023 | |
NM_001160037.1 | NP_001153509.1 | 10 | 4719 | ||
NM_015178.2 | NP_055993.2 | 10 | 5211 | ||
XM_017013251.1 | XP_016868740.1 | 11 | 9538 | ||
GenBank mRNA | AB018260.2 | - | 10 | 5211 | |
AY009093.1 | - | 10 | 4865 | ||
BG255120.1 | - | 1 | 405 |