Gene Transcripts
- Gene Symbol
- SPACA9
- Entrez Gene ID
- 11092
- Gene Name
- sperm acrosome associated 9
- Gene Aliases
- C9orf9, Mast
- Chromosome Location
- Chr. 9: 132878320 - 132890314 on Build GRCh38
- UniGene ID
- Hs.62595
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001316897.1 |
NP_001303826.1
|
3, 4 |
504 |
|
NM_001316898.1 |
NP_001303827.1
|
3, 4 |
497 |
|
NM_001316900.1 |
NP_001303829.1
|
3, 4 |
504 |
|
NM_018956.4 |
NP_061829.3
|
3, 4 |
504 |
|
XM_017014231.1 |
XP_016869720.1
|
3, 4 |
500 |
|
GenBank mRNA |
AJ011375.1 |
- |
3, 4 |
492 |
|
AK075259.1 |
- |
3, 4 |
497 |
|
BC012940.1 |
- |
3, 4 |
454 |
4478288 |
CK906175.1 |
- |
3, 4 |
450 |
|
CR457210.1 |
- |
2, 3 |
343 |
|
DA134573.1 |
- |
3, 4 |
497 |
|
View Full Product Details