Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003328.2 | NP_003319.2 | 6 | 571 | |
XM_011513747.2 | XP_011512049.1 | 5 | 523 | ||
XM_017008580.1 | XP_016864069.1 | 7 | 1773 | ||
XM_017008581.1 | XP_016864070.1 | 5 | 1582 | ||
GenBank mRNA | L27071.1 | - | 6 | 571 |