Gene Transcripts

Gene Symbol
BTG2
Entrez Gene ID
7832
Gene Name
BTG anti-proliferation factor 2
Gene Aliases
APRO1, PC3, TIS21
Chromosome Location
Chr. 1: 203305536 - 203309602 on Build GRCh38
UniGene ID
Hs.519162
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006763.2 NP_006754.1 2 1247
GenBank mRNA BC105948.1 - 2 1220
BC105949.1 - 2 1220 40032675
U72649.1 - 2 1246

View Full Product Details