Gene Transcripts

Gene Symbol
ZNF79
Entrez Gene ID
7633
Gene Name
zinc finger protein 79
Gene Aliases
pT7
Chromosome Location
Chr. 9: 127424374 - 127446289 on Build GRCh38
UniGene ID
Hs.522399
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286696.1 NP_001273625.1 5 978
NM_001286697.1 NP_001273626.1 6 980
NM_001286698.1 NP_001273627.1 3 755
NM_001322260.1 NP_001309189.1 6 1109
NM_007135.2 NP_009066.2 5 871
XM_006717279.3 XP_006717342.1 6 1079
GenBank mRNA AK054606.1 - 5 863
AK302284.1 - 5 920
AK316233.1 - 6 916
BC062309.1 - 5 826 3944657

View Full Product Details