Gene Transcripts

Gene Symbol
ALDH5A1
Entrez Gene ID
7915
Gene Name
aldehyde dehydrogenase 5 family member A1
Gene Aliases
SSADH, SSDH
Chromosome Location
Chr. 6: 24494969 - 24537207 on Build GRCh38
UniGene ID
Hs.371723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080.3 NP_001071.1 7 1137
NM_170740.1 NP_733936.1 8 1176
GenBank mRNA AJ427355.2 - 7 1109
AK290800.1 - 7 1255
AK294699.1 - 8 914
AK314357.1 - 7 1137
AK315380.1 - 7 1232
BC034321.1 - 7 1137 4829533
KJ534766.1 - 7 1052
KJ534767.1 - 7 1109
KJ534768.1 - 7 1109
KJ534769.1 - 8 1148
L34820.1 - 4 473
Y11192.1 - 7 1109

View Full Product Details