Gene Transcripts

Gene Symbol
C17orf53
Entrez Gene ID
78995
Gene Name
chromosome 17 open reading frame 53
Gene Aliases
Chromosome Location
Chr. 17: 44141902 - 44162477 on Build GRCh38
UniGene ID
Hs.437059
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171251.2 NP_001164722.1 7 1847
NM_001321310.1 NP_001308239.1 5 1622
NM_001321311.1 NP_001308240.1 7 1844
NM_024032.4 NP_076937.2 7 1850
XM_011525187.1 XP_011523489.1 7 1697
XM_011525188.1 XP_011523490.1 6 1616
XM_011525189.1 XP_011523491.1 6 1559
XM_011525190.1 XP_011523492.1 6 1556
XM_011525191.2 XP_011523493.1 7 1700
XM_011525193.1 XP_011523495.1 5 1472
XM_011525197.2 XP_011523499.1 4 865
XM_017025031.1 XP_016880520.1 7 1873
XM_017025032.1 XP_016880521.1 7 1869
GenBank mRNA AK291924.1 - 7 1841
AL834295.1 - 7 1794
BC000121.1 - 5 1502 3352851

View Full Product Details