Gene Transcripts

Gene Symbol
CEP112
Entrez Gene ID
201134
Gene Name
centrosomal protein 112
Gene Aliases
CCDC46, MACOCO
Chromosome Location
Chr. 17: 65635540 - 66192131 on Build GRCh38
UniGene ID
Hs.408676
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037325.2 NP_001032402.1 3 597
NM_001199165.2 NP_001186094.1 23 2728
NM_001302891.1 NP_001289820.1 21 2678
XM_005257119.4 XP_005257176.1 23 2822
XM_005257125.3 XP_005257182.1 15 1812
XM_005257126.4 XP_005257183.1 12 1477
XM_006721740.2 XP_006721803.1 23 2694
XM_011524461.2 XP_011522763.1 23 2835
XM_011524462.2 XP_011522764.1 23 2824
XM_011524463.2 XP_011522765.1 23 2824
XM_011524465.2 XP_011522767.1 20 2151
XM_011524466.2 XP_011522768.1 13 1630
XM_011524467.2 XP_011522769.1 12 1472
XM_017024302.1 XP_016879791.1 21 2565
GenBank mRNA AF458591.1 - 13 1635
AK097612.1 - 5 696
AK299075.1 - 23 2679
BC029524.1 - 3 597
BC131805.1 - 21 2506 40119069
BX097629.1 - 3 275

View Full Product Details