Gene Transcripts
- Gene Symbol
- EFCAB13
- Entrez Gene ID
- 124989
- Gene Name
- EF-hand calcium binding domain 13
- Gene Aliases
- C17orf57
- Chromosome Location
- Chr. 17: 47323961 - 47441312 on Build GRCh38
- UniGene ID
- Hs.463303
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152347.4 |
NP_689560.3
|
25 |
3337 |
|
GenBank mRNA |
BC036407.2 |
- |
25 |
3317 |
|
View Full Product Details