Gene Transcripts

Gene Symbol
CCDC178
Entrez Gene ID
374864
Gene Name
coiled-coil domain containing 178
Gene Aliases
C18orf34
Chromosome Location
Chr. 18: 32937402 - 33441040 on Build GRCh38
UniGene ID
Hs.115461
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105528.1 NP_001098998.1 8 657
NM_001308126.1 NP_001295055.1 8 657
NM_198995.2 NP_945346.2 9 726
XM_011525948.1 XP_011524250.1 9 726
XM_011525951.1 XP_011524253.1 9 726
XM_011525954.2 XP_011524256.1 9 726
XM_011525955.1 XP_011524257.1 9 726
XM_017025721.1 XP_016881210.1 9 726
XM_017025722.1 XP_016881211.1 9 747
XM_017025723.1 XP_016881212.1 9 756
XM_017025724.1 XP_016881213.1 3 290
XM_017025725.1 XP_016881214.1 3 907
GenBank mRNA AK126038.1 - 9 726
BC062719.1 - 9 680
BC091517.1 - 9 698 30389491
BC130535.1 - 8 594 40146566
BC144178.1 - 8 594 9052698

View Full Product Details