Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002889.3 | NP_002880.1 | 6 | 713 | |
GenBank mRNA | AB015632.1 | - | 6 | 668 | |
AK092804.1 | - | 4 | 1879 | ||
BC000069.2 | - | 6 | 644 | 3506067 | |
BM919890.1 | - | 5 | 648 | ||
BX448753.2 | - | 6 | 712 | ||
U77594.2 | - | 6 | 656 |