Gene Transcripts

Gene Symbol
ROR1
Entrez Gene ID
4919
Gene Name
receptor tyrosine kinase like orphan receptor 1
Gene Aliases
NTRKR1, dJ537F10.1
Chromosome Location
Chr. 1: 63774019 - 64181498 on Build GRCh38
UniGene ID
Hs.128753
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005012.3 NP_005003.2 9 1927
XM_011541526.1 XP_011539828.1 9 1526
XM_017001376.1 XP_016856865.1 8 1827
XM_017001377.1 XP_016856866.1 11 4470
GenBank mRNA AK309189.1 - 6 1169
BC128386.1 - 10 1537
M97675.1 - 9 1903

View Full Product Details