Gene Transcripts

Gene Symbol
AQP4
Entrez Gene ID
361
Gene Name
aquaporin 4
Gene Aliases
MIWC, WCH4
Chromosome Location
Chr. 18: 26852038 - 26865844 on Build GRCh38
UniGene ID
Hs.315369
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317384.1 NP_001304313.1 2 272
NM_001317387.1 NP_001304316.1 2 272
NM_001650.5 NP_001641.1 2 272
NM_004028.4 NP_004019.1 1 399
XM_011525942.2 XP_011524244.1 2 314
GenBank mRNA AB209156.1 - 2 138
AK222684.1 - 2 184
AK295069.1 - 2 143
BC022286.1 - 2 137 4717755
BI596912.1 - 2 172
BI667387.1 - 2 190
D63412.1 - 2 137
HQ901095.1 - 2 246
KF055862.1 - 2 117
U34845.1 - 1 335
U63622.1 - 2 156
U63623.1 - 1 60

View Full Product Details