Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001731.2 | NP_001722.1 | 2 | 780 | |
GenBank mRNA | AK307057.1 | - | 3 | 533 | |
AK313309.1 | - | 2 | 731 | ||
BC016759.2 | - | 2 | 722 | 2987983 | |
BC064953.1 | - | 2 | 723 | 6050290 | |
BT019839.1 | - | 2 | 418 | ||
BT019840.1 | - | 2 | 418 | ||
BT019841.1 | - | 2 | 418 | ||
CR456876.1 | - | 2 | 418 | ||
X61123.1 | - | 2 | 726 |