Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145296.1 | NP_660339.1 | 5 | 602 | |
XM_005258620.2 | XP_005258677.1 | 5 | 584 | ||
XM_017026452.1 | XP_016881941.1 | 5 | 511 | ||
GenBank mRNA | AF363368.1 | - | 5 | 602 | |
AK313028.1 | - | 5 | 584 | ||
BC068457.1 | - | 5 | 583 | ||
BC113963.1 | - | 2 | 175 |