Gene Transcripts

Gene Symbol
LRRC75A
Entrez Gene ID
388341
Gene Name
leucine rich repeat containing 75A
Gene Aliases
C17orf76, FAM211A
Chromosome Location
Chr. 17: 16442005 - 16492191 on Build GRCh38
UniGene ID
Hs.25425
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113567.2 NP_001107039.1 4 1699
NM_207387.3 NP_997270.2 3 1583
XM_011523845.2 XP_011522147.1 5 1772
XM_017024619.1 XP_016880108.1 4 1468
XM_017024620.1 XP_016880109.1 5 1631
GenBank mRNA AK093015.1 - 3 1242
AL833245.1 - 3 1609
AL833260.1 - 3 1609

View Full Product Details