Gene Transcripts

Gene Symbol
LCNL1
Entrez Gene ID
401562
Gene Name
lipocalin like 1
Gene Aliases
Chromosome Location
Chr. 9: 136982993 - 136985758 on Build GRCh38
UniGene ID
Hs.695210
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK127236.1 - 1 1904
BC042845.1 - 1 484
BC137559.1 - 1 1393 9021559
BC137567.1 - 1 1393 9021567

View Full Product Details