Gene Transcripts
- Gene Symbol
- Ptma
- Entrez Gene ID
- 19231
- Gene Name
- prothymosin alpha
- Gene Aliases
- Thym
- Chromosome Location
- Chr. 1: 86526736 - 86530698 on Build GRCm38
- UniGene ID
- Mm.19187
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X56135.1 |
- |
5 |
505 |
|
View Full Product Details