Gene Transcripts

Gene Symbol
Aldh1a2
Entrez Gene ID
19378
Gene Name
aldehyde dehydrogenase family 1, subfamily A2
Gene Aliases
AV116159, Aldh1a7, Raldh1, Raldh2
Chromosome Location
Chr. 9: 71215789 - 71296243 on Build GRCm38
UniGene ID
Mm.42016
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009022.4 NP_033048.2 8 931
GenBank mRNA AK078553.1 - 8 872
BC075704.1 - 8 898 30471325
X99273.1 - 8 876

View Full Product Details