Gene Transcripts
- Gene Symbol
- Mapk11
- Entrez Gene ID
- 19094
- Gene Name
- mitogen-activated protein kinase 11
- Gene Aliases
- P38b, Prkm11, Sapk2, Sapk2b, p38-2, p38beta, p38beta2
- Chromosome Location
- Chr. 15: 89142481 - 89149623 on Build GRCm38
- UniGene ID
- Mm.91969
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC064737.1 |
- |
11 |
901 |
|
View Full Product Details