Gene Transcripts

Gene Symbol
Ankrd13b
Entrez Gene ID
268445
Gene Name
ankyrin repeat domain 13b
Gene Aliases
AW124583, B930093C12Rik
Chromosome Location
Chr. 11: 77470485 - 77489678 on Build GRCm38
UniGene ID
Mm.39296
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172945.2 NP_766533.2 16 2846
GenBank mRNA AK081149.1 - 1976
BC022693.1 - 5 1392
BC039786.1 - 5 1810
BC056928.1 - 9 2332
BC071189.1 - 10 1932
BC098207.1 - 7 2142

View Full Product Details