Gene Transcripts

Gene Symbol
Mcpt2
Entrez Gene ID
17225
Gene Name
mast cell protease 2
Gene Aliases
MMCP-2, Mcp-2
Chromosome Location
Chr. 14: 56021584 - 56044635 on Build GRCm38
UniGene ID
Mm.4409
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008571.1 NP_032597.1 4 567
GenBank mRNA AK133521.1 - 4 568
AY007570.1 - 4 567
BC119248.1 - 4 560 8733997
BC119250.1 - 4 560 8733999

View Full Product Details