Gene Transcripts

Gene Symbol
Pcnt
Entrez Gene ID
18541
Gene Name
pericentrin (kendrin)
Gene Aliases
AW476095, C86676, KEN, Pcnt2, kendrin, m239Asp, m275Asp
Chromosome Location
Chr. 10: 76351254 - 76444122 on Build GRCm38
UniGene ID
Mm.251794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282992.1 NP_001269921.1 5 908
NM_008787.3 NP_032813.3 5 908
GenBank mRNA AB207233.1 - 5 782
AK145315.1 - 6 729
BC085244.1 - 5 732
BC100380.1 - 5 703
BU518252.1 - 429

View Full Product Details