Gene Transcripts

Gene Symbol
Myo5a
Entrez Gene ID
17918
Gene Name
myosin VA
Gene Aliases
9630007J19Rik, AI413174, AI661011, Dbv, MVa, Myo5, MyoVA, Sev-1, d, d-120J, flail, flr
Chromosome Location
Chr. 9: 75069298 - 75223688 on Build GRCm38
UniGene ID
Mm.3645
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010864.2 NP_034994.2 3 681
GenBank mRNA AK032713.1 - 3 682
AK149701.1 - 1 125
X57377.1 - 3 294

View Full Product Details