Gene Transcripts

Gene Symbol
Aktip
Entrez Gene ID
14339
Gene Name
thymoma viral proto-oncogene 1 interacting protein
Gene Aliases
AL023020, Fif, Ft, Ft1, Fts
Chromosome Location
Chr. 8: 91109871 - 91135494 on Build GRCm38
UniGene ID
Mm.324588
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK017861.1 - 5 552
AK146459.1 - 6 668
AK158704.1 - 6 670
BC008130.1 - 7 673 3498984
BC034861.1 - 6 677
X71978.1 - 6 692
Z67963.1 - 6 692

View Full Product Details