Gene Transcripts

Gene Symbol
Ap3d1
Entrez Gene ID
11776
Gene Name
adaptor-related protein complex 3, delta 1 subunit
Gene Aliases
AA407035, Ap3d, Bolvr, mBLVR1, mh, mocha
Chromosome Location
Chr. 10: 80706956 - 80742282 on Build GRCm38
UniGene ID
Mm.209294
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007460.1 NP_031486.1 5, 6 686
GenBank mRNA AB004305.1 - 5, 6 686
AK141789.1 - 5, 6 534
AK155518.1 - 5, 6 736
AK155825.1 - 5, 6 713
BC022171.1 - 5, 6 691
BC037477.1 - 5, 6 687
BC048786.1 - 5, 6 676 6489789
BC053066.1 - 5, 6 702 5704929

View Full Product Details