Gene Transcripts
- Gene Symbol
- Akap2
- Entrez Gene ID
- 11641
- Gene Name
- A kinase (PRKA) anchor protein 2
- Gene Aliases
- AA959716, AI649048, Akap-2, Akap-kl, B230340M18Rik, Prka2
- Chromosome Location
- Chr. 4: 57845248 - 57896984 on Build GRCm38
- UniGene ID
- Mm.467752
- Species
- Mus musculus
- Gene Symbol
- Pakap
- Entrez Gene ID
- 677884
- Gene Name
- paralemmin A kinase anchor protein
- Gene Aliases
- AF064781, Gm20459, Palm2-Akap2, Palm2Akap2, Palm2akap2
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.331630
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK077020.1 |
- |
3 |
2014 |
|
View Full Product Details