Gene Transcripts

Gene Symbol
Aldh3b1
Entrez Gene ID
67689
Gene Name
aldehyde dehydrogenase 3 family, member B1
Gene Aliases
1700001N19Rik, ALDH4, ALDH7
Chromosome Location
Chr. 19: 3913491 - 3930817 on Build GRCm38
UniGene ID
Mm.109341
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026316.2 NP_080592.2 6, 7 592
GenBank mRNA AF362571.1 - 5, 6 514
AK154500.1 - 6, 7 638
AK154723.1 - 6, 7 685
AK159447.1 - 6, 7 606
AK160106.1 - 6, 7 609
BC023305.1 - 6, 7 613
BC046597.1 - 6, 7 592 6312211

View Full Product Details