Gene Transcripts

Gene Symbol
Lrrk1
Entrez Gene ID
233328
Gene Name
leucine-rich repeat kinase 1
Gene Aliases
AW319595, C230002E15Rik, D130026O16Rik, mKIAA1790
Chromosome Location
Chr. 7: 66258745 - 66388341 on Build GRCm38
UniGene ID
Mm.33284
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146191.3 NP_666303.3 29, 30 4919
GenBank mRNA AK076432.1 - 2, 3 246
AK147469.1 - 29, 30 4905

View Full Product Details