Gene Transcripts
- Gene Symbol
- Atad3a
- Entrez Gene ID
- 108888
- Gene Name
- ATPase family, AAA domain containing 3A
- Gene Aliases
- 2400004H09Rik, Atad3, Tob3, mKIAA1273
- Chromosome Location
- Chr. 4: 155740640 - 155761108 on Build GRCm38
- UniGene ID
- Mm.241152
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_179203.3 |
NP_849534.2
|
12, 13 |
1358 |
|
GenBank mRNA |
AF343079.1 |
- |
12, 13 |
1404 |
|
AK075921.1 |
- |
12, 13 |
1354 |
|
AK148974.1 |
- |
12, 13 |
1357 |
|
AK149702.1 |
- |
12, 13 |
1357 |
|
AK150037.1 |
- |
12, 13 |
1357 |
|
AK165724.1 |
- |
12, 13 |
1356 |
|
AK166099.1 |
- |
12, 13 |
1354 |
|
AK172038.1 |
- |
12, 13 |
1354 |
|
AK172350.1 |
- |
12, 13 |
1359 |
|
AK218475.1 |
- |
2, 3 |
111 |
|
BC023301.1 |
- |
12, 13 |
1295 |
3671787 |
BC058373.1 |
- |
12, 13 |
1342 |
6828037 |
BC060036.1 |
- |
12, 13 |
1290 |
6530155 |
View Full Product Details