Gene Transcripts

Gene Symbol
Bloc1s3
Entrez Gene ID
232946
Gene Name
biogenesis of lysosomal organelles complex-1, subunit 3
Gene Aliases
BC043666, Blos3, E230011O18, rp
Chromosome Location
Chr. 7: 19505804 - 19508371 on Build GRCm38
UniGene ID
Mm.64557
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177692.3 NP_808360.2 2 1156
GenBank mRNA AK054006.1 - 2 1175
AK153811.1 - 1194
BC025913.1 - 1 946
BC043666.1 - 2 1156 5025443

View Full Product Details