Gene Transcripts

Gene Symbol
C1ql3
Entrez Gene ID
227580
Gene Name
C1q-like 3
Gene Aliases
1110065A22Rik, AI661623, C1ql, C1qtnf13, CTRP13, K100
Chromosome Location
Chr. 2: 13001887 - 13010864 on Build GRCm38
UniGene ID
Mm.229322
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153155.2 NP_694795.1 2 981
GenBank mRNA AB044560.1 - 1864

View Full Product Details