Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173403.2 | NP_775579.2 | 3 | 1368 | |
GenBank mRNA | AK087479.1 | - | 3 | 1369 | |
AK149202.1 | - | 1369 | |||
BC132529.1 | - | 3 | 1280 | 40130806 | |
BC138048.1 | - | 3 | 1280 | 8861067 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001024147.2 | NP_001019318.1 | 3 | 698 | |
GenBank mRNA | AK084047.1 | - | 3 | 699 |