Gene Transcripts

Gene Symbol
Scgb2b24
Entrez Gene ID
233090
Gene Name
secretoglobin, family 2B, member 24
Gene Aliases
Abpbg24, Abpz, C2b, Scgb2b3
Chromosome Location
Chr. 7: 33737193 - 33739295 on Build GRCm38
UniGene ID
Mm.261648
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177446.2 NP_803229.1 2, 3 250
GenBank mRNA AA794526.1 - 2, 3 250
AY370635.1 - 2, 3 267
BC115602.1 - 2, 3 250 40102624
BC115603.1 - 2, 3 250 40102625
GU269236.1 - 2, 3 240

View Full Product Details